missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(1-adamantyl)benzimidazolium chloride, 95%, Thermo Scientific™

Catalog Number p-4258765
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy
852634-41-4
C27H35N2·Cl
423.05
GMYUUHXEUIPSKA-UHFFFAOYSA-M
1,3-bis 1-adamanthyl benzimidazolium chloride, 1,3-bis 1-adamantyl benzimidazolium chloride, 1,3-bis 1-adamantyl-benzimidazolium chloride, 1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride, 1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride, 1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride
16218257
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
This item is not returnable. View return policy
852634-41-4
C27H35N2·Cl
423.05
GMYUUHXEUIPSKA-UHFFFAOYSA-M
1,3-bis 1-adamanthyl benzimidazolium chloride, 1,3-bis 1-adamantyl benzimidazolium chloride, 1,3-bis 1-adamantyl-benzimidazolium chloride, 1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride, 1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride, 1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride
16218257
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
CAS Min % 94.0
CAS Max % 100.0
Melting Point 283.9°C to 285.6°C
Color Gray to White
Elemental Analysis 72.06 to 81.26% C, 6.22 to 7.02% N
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 500 mg
Formula Weight 423.05
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1, 3-Bis(1-Adamantyl)benzimidazolium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Bis(1-adamantyl)-benzimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'