missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(1-adamantyl)benzimidazolium chloride, 95%, ACROS Organics™

Brand:  Acros Organics 435070010

Code : 29

Additional Details : CAS Number : 852634-41-4 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11966651

  • 138.00 € / 1g

Estimated Shipment
28-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

852634-41-4
423.05
1,3-bis 1-adamanthyl benzimidazolium chloride, 1,3-bis 1-adamantyl benzimidazolium chloride, 1,3-bis 1-adamantyl-benzimidazolium chloride, 1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride, 1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride, 1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
C27H35N2·Cl
GMYUUHXEUIPSKA-UHFFFAOYSA-M
16218257
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]

Specifications

1, 3-Bis(1-Adamantyl)benzimidazolium chloride
Authentic
Gray to White
1g
94.0
94% min. (HPLC)
1,3-bis 1-adamanthyl benzimidazolium chloride, 1,3-bis 1-adamantyl benzimidazolium chloride, 1,3-bis 1-adamantyl-benzimidazolium chloride, 1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride, 1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride, 1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
423.05
423.05
95%
72.06 to 81.26% C, 6.22 to 7.02% N
Glass Bottle
283.9°C to 285.6°C
852634-41-4
100.0
C27H35N2·Cl
GMYUUHXEUIPSKA-UHFFFAOYSA-M
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
16218257
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Bis(1-adamantyl)-benzimidazolium chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents