missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-96188
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
504-01-8
C6H12O2
116.16
MFCD00039458
RLMGYIOTPQVQJR-UHFFFAOYSA-N
1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
10433
cyclohexane-1,3-diol
C1CC(CC(C1)O)O
This item is not returnable. View return policy
504-01-8
C6H12O2
116.16
MFCD00039458
RLMGYIOTPQVQJR-UHFFFAOYSA-N
1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
10433
cyclohexane-1,3-diol
C1CC(CC(C1)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 30°C
Color Yellow
Boiling Point 246°C to 247°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(OH)2
Refractive Index 1.4950 to 1.497
Quantity 10 g
Beilstein 06, 740
Formula Weight 116.16
Percent Purity 98%
Physical Form Viscous Liquid After Melting
Chemical Name or Material 1, 3-Cyclohexanediol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'