missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole, 97%, Thermo Scientific™

Catalog Number p-2694945
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
58419-69-5
C9H8BrN3
238.088
MFCD04113600
NOUAEJFAVOHNSY-UHFFFAOYSA-N
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole
2776489
1-[4-(bromomethyl)phenyl]-1,2,4-triazole
C1=CC(=CC=C1CBr)N2C=NC=N2
This item is not returnable. View return policy
58419-69-5
C9H8BrN3
238.088
MFCD04113600
NOUAEJFAVOHNSY-UHFFFAOYSA-N
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole
2776489
1-[4-(bromomethyl)phenyl]-1,2,4-triazole
C1=CC(=CC=C1CBr)N2C=NC=N2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 319
Percent Purity 97%
Chemical Name or Material 1-[4-(Bromomethyl)phenyl]-1H-1, 2, 4-triazole hydrobromide hydrate
compliance-icons
Product Identifier
  • 1-¢4-(Bromomethyl)phenyl!-1H-1,2,4-triazole hydrobromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'