missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Maybridge

78.40 € - 156.80 €

Chemical Identifiers

CAS 98534-80-6
Molecular Formula C11H6ClF3N2O2
Molecular Weight (g/mol) 290.626
MDL Number MFCD00068142
InChI Key HSZJMDBOSZVDOK-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl-2h-pyrazole-4-carboxylic acid, maybridge1_004008, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4, 1-4-chlorophenyl-5-trifluoromethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl
PubChem CID 688687
IUPAC Name 1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
SMILES C1=CC(=CC=C1N2C(=C(C=N2)C(=O)O)C(F)(F)F)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10005962 SDS View Documents Maybridge
GK01550EA
10g Amber Glass Bottle 78.40 €
10g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 07-05-2020
Please to purchase this item. Need a web account? Register with us today!
10662705 SDS View Documents Maybridge
GK01550EB
25g Amber Glass Bottle 156.80 €
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 07-05-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

98534-80-6
290.626
HSZJMDBOSZVDOK-UHFFFAOYSA-N
688687
C1=CC(=CC=C1N2C(=C(C=N2)C(=O)O)C(F)(F)F)Cl
C11H6ClF3N2O2
MFCD00068142
1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl-2h-pyrazole-4-carboxylic acid, maybridge1_004008, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4, 1-4-chlorophenyl-5-trifluoromethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl
1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid

Specifications

1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
10g
96.5
97%
MFCD00068142
HSZJMDBOSZVDOK-UHFFFAOYSA-N
1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
688687
97%
Amber Glass Bottle
98534-80-6
100.0
C11H6ClF3N2O2
1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl-2h-pyrazole-4-carboxylic acid, maybridge1_004008, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4, 1-4-chlorophenyl-5-trifluoromethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl
C1=CC(=CC=C1N2C(=C(C=N2)C(=O)O)C(F)(F)F)Cl
290.626
290.63