missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals

Catalog Number p-101029
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
280-57-9
C6H12N2
112.17
MFCD00006689
IMNIMPAHZVJRPE-UHFFFAOYSA-N
1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
9237
1,4-diazabicyclo[2.2.2]octane
C1CN2CCN1CC2
This item is not returnable. View return policy
280-57-9
C6H12N2
112.17
MFCD00006689
IMNIMPAHZVJRPE-UHFFFAOYSA-N
1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
9237
1,4-diazabicyclo[2.2.2]octane
C1CN2CCN1CC2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 160.0°C
Color White to Yellow
Density 1.1400g/mL
Boiling Point 174.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 25 g
Fieser 01,1203; 02,99; 04,119; 05,176; 06,157; 07,86; 11,153; 12,155; 13,92; 15,109
Merck Index 15,9834
Specific Gravity 1.14
Solubility Information Solubility in water: 46g/100mL (26°C). Other solubilities: soluble in most common organic solvents
Sublimation Point 20 °C
Formula Weight 112.17
Percent Purity 97%
Physical Form Hygroscopic Crystals
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Diazabicyclo¢2.2.2!octane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'