missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[3.2.2]nonane, 95%, ACROS Organics™

Brand:  Acros Organics 438900050

Code : 29

Additional Details : CAS Number : 283-38-5 Weight : 0.05500kg Transport : UN number : 3259 Chem class : 8 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11946681

  • 688.00 € / 5g

Estimated Shipment
26-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

283-38-5
126.2
1,4-diazabicyclo 3.2.2 nonane, 1,4-diazobicylco 3.2.2 nonane, 1,4-diaza-bicyclo 3.2.2 nonane, 1,4-diazabicyclo-3.2.2 nonane, 1,4-diazabicyclo 3.2.2 _nonane, 1,4-diazabicyclo 3.2.2-nonane
1,4-diazabicyclo[3.2.2]nonane
C7H14N2
XJKNACDCUAFDHD-UHFFFAOYSA-N
398407
C1CN2CCC1NCC2

Specifications

1,4-Diazabicyclo[3.2.2]nonane
Glass bottle
Colorless to Yellow
5g
94.0
95%
1,4-diazabicyclo 3.2.2 nonane, 1,4-diazobicylco 3.2.2 nonane, 1,4-diaza-bicyclo 3.2.2 nonane, 1,4-diazabicyclo-3.2.2 nonane, 1,4-diazabicyclo 3.2.2 _nonane, 1,4-diazabicyclo 3.2.2-nonane
C1CN2CCC1NCC2
126.2
126.2
95%
Authentic
77.0°C to 79.0°C (6.0 Torr)
58.0°C to 79.0°C
283-38-5
100.0
C7H14N2
XJKNACDCUAFDHD-UHFFFAOYSA-N
1,4-diazabicyclo[3.2.2]nonane
398407
Waxy Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,4-Diazabicyclo¢3.2.2!nonane

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents