missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[3.2.2]nonane, 95%, Thermo Scientific Chemicals

Catalog Number p-4228580
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
283-38-5
C7H14N2
126.2
XJKNACDCUAFDHD-UHFFFAOYSA-N
1,4-diazabicyclo 3.2.2 nonane, 1,4-diazobicylco 3.2.2 nonane, 1,4-diaza-bicyclo 3.2.2 nonane, 1,4-diazabicyclo-3.2.2 nonane, 1,4-diazabicyclo 3.2.2 _nonane, 1,4-diazabicyclo 3.2.2-nonane
398407
1,4-diazabicyclo[3.2.2]nonane
C1CN2CCC1NCC2
This item is not returnable. View return policy
283-38-5
C7H14N2
126.2
XJKNACDCUAFDHD-UHFFFAOYSA-N
1,4-diazabicyclo 3.2.2 nonane, 1,4-diazobicylco 3.2.2 nonane, 1,4-diaza-bicyclo 3.2.2 nonane, 1,4-diazabicyclo-3.2.2 nonane, 1,4-diazabicyclo 3.2.2 _nonane, 1,4-diazabicyclo 3.2.2-nonane
398407
1,4-diazabicyclo[3.2.2]nonane
C1CN2CCC1NCC2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 58.0°C to 79.0°C
Color Colorless to Yellow
Boiling Point 77.0°C to 79.0°C (6.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 126.2
Percent Purity 95%
Physical Form Waxy Solid
Chemical Name or Material 1,4-Diazabicyclo[3.2.2]nonane
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diazabicyclo¢3.2.2!nonane
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'