missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromo-2-nitrobenzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as an application intermediate. Some of the other applications include HPLC, NMR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as an application intermediate. Some of the other applications include HPLC, NMR.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Strong bases.
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as an application intermediate. Some of the other applications include HPLC, NMR.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Strong bases.
Specifications
Melting Point | 82°C to 85°C |
Density | 2.374 |
Quantity | 100 g |
Beilstein | 1950425 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 280.91 |
Percent Purity | 99% |
Chemical Name or Material | 1,4-Dibromo-2-nitrobenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'