missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™
111.00 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT
Specifications
pH | 6 to 8 |
Melting Point | 12°C |
Color | Colorless |
Boiling Point | 101°C |
Quantity | 1 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 41mbar at 20°C |
Viscosity | 1.32 mPaS at 20°C |
Formula Weight | 88.11g/mol |
Safety and Handling
Product Identifier
- 1,4-Dioxane RPE
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'