missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluoro-2-methylphenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61278.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416416

  • 74.73 € / 1g


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluoro-2-methylphenyl)-3-[4-(trifluoromethyl)phenyl]urea
MFCD26143280

Specifications

1-(4-Fluoro-2-methylphenyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
MFCD26143280
Solid
White
97%
312.26
Videos
SDS