missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadiene, 99+%, stabilized, purified by redistillation, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4323168
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
111-78-4
C8H12
108.18
MFCD00001752
VYXHVRARDIDEHS-QGTKBVGQSA-N
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
10937607
(5Z)-cycloocta-1,5-diene
C1C\C=C/CC\C=C/1
111-78-4
C8H12
108.18
MFCD00001752
VYXHVRARDIDEHS-QGTKBVGQSA-N
1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
10937607
(5Z)-cycloocta-1,5-diene
C1C\C=C/CC\C=C/1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -69.5°C
Color Colorless
Density 0.8800g/mL
Boiling Point 151°C (101.3 kPa)
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4920 to 1.4940
Quantity 100 mL
Beilstein 05,116
Merck Index 15,273
Specific Gravity 0.88
Solubility Information Solubility in water: 780mg/l (20°C). Other solubilities: soluble in benzene and carbon tetrachloride
Stabilizer 100 to 200ppm octadecyl 3-(3«, 5«-di-tert-butyl-4«-hydroxyphenyl)propionate
Viscosity 1.3 mPa.s (20°C)
Formula Weight 108.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 5-Cyclooctadiene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'