missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021746
Click to view available options
Quantity:
100 g
500 g
2500 g
124-09-4
C6H16N2
MFCD00008243
NAQMVNRVTILPCV-UHFFFAOYSA-N
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
16402
CHEBI:39618
hexane-1,6-diamine
124-09-4
C6H16N2
MFCD00008243
NAQMVNRVTILPCV-UHFFFAOYSA-N
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
16402
CHEBI:39618
hexane-1,6-diamine

1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

Solubility
Soluble in water. Slightly soluble in ether, alcohol and benzene.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents, acid chlorides, carbon dioxide and acid anhydrides.
TRUSTED_SUSTAINABILITY
Melting Point 39°C to 43°C
Density 0.84
pH 12.4
Boiling Point 199°C to 204°C
Flash Point 85°C (185°F)
Odor Amine-like
Linear Formula H2N(CH2)6NH2
Quantity 100 g
UN Number UN2280
Beilstein 1098307
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,4695
Solubility Information Soluble in water. Slightly soluble in ether,alcohol and benzene.
SMILES C(CCCN)CCN
Molecular Weight (g/mol) 116.208
Formula Weight 116.21
Percent Purity ≥98%
Chemical Name or Material 1,6-Diaminohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'