missing translation for 'onlineSavingsMsg'
Learn More

1,8-Nonadiyne, 97%, Thermo Scientific Chemicals

Catalog Number p-7027207
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2396-65-8
C9H12
120.195
MFCD00008581
DMOVPHYFYSASTC-UHFFFAOYSA-N
1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci
16969
nona-1,8-diyne
C#CCCCCCC#C
This item is not returnable. View return policy
2396-65-8
C9H12
120.195
MFCD00008581
DMOVPHYFYSASTC-UHFFFAOYSA-N
1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci
16969
nona-1,8-diyne
C#CCCCCCC#C

It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from heavy metal powders and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY
Melting Point -21°C
Density 0.799
Boiling Point 55°C to 56°C (13 mmHg)
Flash Point 41°C (105°F)
Odor Characteristic
Linear Formula HC≡C(CH2)5C≡CH
Refractive Index 1.449
Quantity 5 g
UN Number UN3295
Beilstein 1733443
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.2
Percent Purity 97%
Chemical Name or Material 1,8-Nonadiyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'