missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,9-Nonanediol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Specifications
Melting Point | 45°C to 48°C |
Density | 0.95 |
Boiling Point | 288°C |
Flash Point | 162°C (323°F) |
Quantity | 25 g |
Beilstein | 1737531 |
Solubility Information | Soluble in methanol. |
Formula Weight | 160.26 |
Percent Purity | 97% |
Chemical Name or Material | 1,9-Nonanediol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'