missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanediol, 99%, Thermo Scientific Chemicals

Catalog Number p-175484
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3937-56-2
C9H20O2
160.26
MFCD00002991
ALVZNPYWJMLXKV-UHFFFAOYSA-N
1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x
19835
nonane-1,9-diol
C(CCCCO)CCCCO
This item is not returnable. View return policy
3937-56-2
C9H20O2
160.26
MFCD00002991
ALVZNPYWJMLXKV-UHFFFAOYSA-N
1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x
19835
nonane-1,9-diol
C(CCCCO)CCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 44°C to 48°C
Color White
Density 0.9180g/mL
Boiling Point 177°C (15.0 mmHg)
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)9OH
Quantity 25 g
Beilstein 01, 493
Specific Gravity 0.918
Solubility Information Solubility in water: 9g/L (20°C)
Viscosity 52.6 mPa.s (50°C)
Formula Weight 160.26
Percent Purity 99%
Physical Form Fused Crystalline Solid
Chemical Name or Material 1, 9-Nonanediol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'