missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrroline, 97%, Thermo Scientific Chemicals

Catalog Number p-4229085
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6913-92-4
C11H13N
159.23
LRFHKHHUKGZIGE-UHFFFAOYSA-N
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
561506
1-benzyl-2,5-dihydropyrrole
C1C=CCN1CC2=CC=CC=C2
This item is not returnable. View return policy
6913-92-4
C11H13N
159.23
LRFHKHHUKGZIGE-UHFFFAOYSA-N
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
561506
1-benzyl-2,5-dihydropyrrole
C1C=CCN1CC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8370g/mL
Boiling Point 92°C (1.0 hPa)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 0.837
Formula Weight 159.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-pyrroline
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzyl-3-pyrroline
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'