missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2694847
Click to view available options
Quantity:
1 g
100 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10314-98-4
C14H17NO4
263.293
MFCD01568759
URTPNQRAHXRPMP-UHFFFAOYSA-N
1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
736489
1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
10314-98-4
C14H17NO4
263.293
MFCD01568759
URTPNQRAHXRPMP-UHFFFAOYSA-N
1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
736489
1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 263.29
Percent Purity 97%
Chemical Name or Material 1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'