missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-2-(aminomethyl)pyrrolidine, tech. 85%, Thermo Scientific™

Catalog Number 15467745
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
177911-87-4
C10H20N2O2
200.282
MFCD01317793
SOGXYCNKQQJEED-UHFFFAOYSA-N
tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, 2-aminomethyl-1-n-boc-pyrrolidine, +/--2-aminomethyl-1-n-boc-pyrrolidine, 1-boc-2-aminomethyl-pyrrolidine, 1-boc-2-aminomethyl pyrrolidine, 1-n-boc-2-aminomethyl pyrrolidine, 2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-aminomethyl pyrrolidine, 1-boc-2-aminomethylpyrrolidine
2756484
tert-butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1CN
This item is not returnable. View return policy
177911-87-4
C10H20N2O2
200.282
MFCD01317793
SOGXYCNKQQJEED-UHFFFAOYSA-N
tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, 2-aminomethyl-1-n-boc-pyrrolidine, +/--2-aminomethyl-1-n-boc-pyrrolidine, 1-boc-2-aminomethyl-pyrrolidine, 1-boc-2-aminomethyl pyrrolidine, 1-n-boc-2-aminomethyl pyrrolidine, 2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-aminomethyl pyrrolidine, 1-boc-2-aminomethylpyrrolidine
2756484
tert-butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 200.28
Percent Purity 85%
Grade Technical
Chemical Name or Material (+/-)-1-Boc-2-(aminomethyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'