Learn More
1-BOC-3-Aminopiperidine, 97%, ACROS Organics™
Brand: Acros Organics 436600050
Code : 29
Additional Details : CAS Number : 144243-24-3 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
144243-24-3 | |
200.28 | |
1-boc-3-aminopiperidine, n-boc-3-aminopiperidine, 3-amino-1-n-boc-piperidine, +/--3-amino-1-n-boc-piperidine, 3-amino-1-boc-piperidine, 1-n-boc-3-aminopiperidine, 3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3-amino-1-tert-butoxycarbonylpiperidine, 3-aminopiperidine-1-carboxylic acid tert-butyl ester | |
tert-butyl 3-aminopiperidine-1-carboxylate |
C10H20N2O2 | |
AKQXKEBCONUWCL-UHFFFAOYSA-N | |
545809 | |
CC(C)(C)OC(=O)N1CCCC(C1)N |
Specifications
1-BOC-3-Aminopiperidine | |
Colorless to Yellow | |
144243-24-3 | |
100.0 | |
C10H20N2O2 | |
AKQXKEBCONUWCL-UHFFFAOYSA-N | |
tert-butyl 3-aminopiperidine-1-carboxylate | |
545809 | |
Liquid |
Authentic | |
5g | |
96.0 | |
96% min. (GC) | |
1-boc-3-aminopiperidine, n-boc-3-aminopiperidine, 3-amino-1-n-boc-piperidine, +/--3-amino-1-n-boc-piperidine, 3-amino-1-boc-piperidine, 1-n-boc-3-aminopiperidine, 3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3-amino-1-tert-butoxycarbonylpiperidine, 3-aminopiperidine-1-carboxylic acid tert-butyl ester | |
CC(C)(C)OC(=O)N1CCCC(C1)N | |
200.28 | |
200.28 | |
97% |
Safety and Handling
- 1-BOC-3-Aminopiperidine, 97%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.