Learn More
1-Bromo-3,4-difluorobenzene, 98%, ACROS Organics™
Brand: Acros Organics 233190010
Code : 29
Additional Details : CAS Number : 348-61-8 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
348-61-8 | |
193 | |
YMQPKONILWWJQG-UHFFFAOYSA-N | |
67675 | |
C1=CC(=C(C=C1Br)F)F |
C6H3BrF2 | |
MFCD00000304 | |
1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide | |
4-bromo-1,2-difluorobenzene |
Specifications
1-Bromo-3, 4-difluorobenzene | |
33°C | |
Glass bottle | |
1.707 | |
Colorless to Yellow | |
1g | |
348-61-8 | |
100.0 | |
C6H3BrF2 | |
MFCD00000304 | |
Solubility in water: insoluble | |
C1=CC(=C(C=C1Br)F)F | |
193 | |
193 | |
98% |
1.7070g/mL | |
Authentic | |
1.5040 to 1.506 | |
150°C to 151°C | |
-4°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
BrC6H3F2 | |
1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide | |
YMQPKONILWWJQG-UHFFFAOYSA-N | |
4-bromo-1,2-difluorobenzene | |
67675 | |
Liquid |
Safety and Handling
- 1-Bromo-3,4-difluorobenzene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.