missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoadamantane, 99%, Thermo Scientific Chemicals

Catalog Number 10001570
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
768-90-1
C10H15Br
215.13
MFCD00074721
VQHPRVYDKRESCL-UHFFFAOYSA-N
1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane
79106
1-bromoadamantane
C1C2CC3CC1CC(C2)(C3)Br
This item is not returnable. View return policy
768-90-1
C10H15Br
215.13
MFCD00074721
VQHPRVYDKRESCL-UHFFFAOYSA-N
1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane
79106
1-bromoadamantane
C1C2CC3CC1CC(C2)(C3)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 117.0°C to 120.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,chloroform,benzene,,methanol and ethanol
Formula Weight 215.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1-Bromoadamantane, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'