Learn More
1-Bromobenzyl methyl ether, 98%, ACROS Organics™
Brand: Acros Organics 393400250
Code : 29
Additional Details : CAS Number : 52711-30-5 Weight : 0.07500kg
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
52711-30-5 | |
201.06 | |
QFAZLCRHLRJNAW-UHFFFAOYSA-N | |
12927162 | |
COCC1=CC=CC=C1Br |
C8H9BrO | |
MFCD03790887 | |
1-bromo-2-methoxymethyl benzene, 2-bromobenzyl methyl ether, acmc-20ag76, methyl 2-bromobenzyl ether, 1-methoxymethyl-2-bromobenzene, 2-bromobenzyl alcohol, methyl ether, benzene,1-bromo-2-methoxymethyl | |
1-bromo-2-(methoxymethyl)benzene |
Specifications
1-Bromobenzyl methyl ether | |
Glass bottle | |
Colorless to Yellow | |
52711-30-5 | |
100.0 | |
C8H9BrO | |
MFCD03790887 | |
QFAZLCRHLRJNAW-UHFFFAOYSA-N | |
1-bromo-2-(methoxymethyl)benzene | |
12927162 | |
Liquid |
Authentic | |
1.5470 to 1.5490 (20°C, 589nm) | |
25g | |
97.5 | |
97.5% min. (GC) | |
BrC6H4CH2OCH3 | |
1-bromo-2-methoxymethyl benzene, 2-bromobenzyl methyl ether, acmc-20ag76, methyl 2-bromobenzyl ether, 1-methoxymethyl-2-bromobenzene, 2-bromobenzyl alcohol, methyl ether, benzene,1-bromo-2-methoxymethyl | |
COCC1=CC=CC=C1Br | |
201.06 | |
201.06 | |
98% |
Safety and Handling
- 1-Bromobenzyl methyl ether
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.