missing translation for 'onlineSavingsMsg'
Learn More

1-(bromomethyl)-2-phenoxybenzene, 90%, Maybridge

Brand:  Maybridge CC49908DA

Code : 31

Additional Details : CAS Number : 82657-72-5 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 10428413

  • 196.00 € / 1g


Estimated Shipment
29-03-2019
Add to basket

Specifications

Chemical Identifiers

82657-72-5
263.134
1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
1-(bromomethyl)-2-phenoxybenzene
C13H11BrO
YQRIQBOWLXRKKG-UHFFFAOYSA-N
22675469
C1=CC=C(C=C1)OC2=CC=CC=C2CBr

Specifications

1-(Bromomethyl)-2-phenoxybenzene
1g
97%
1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
C1=CC=C(C=C1)OC2=CC=CC=C2CBr
263.134
263.13
Amber glass bottle
82657-72-5
C13H11BrO
YQRIQBOWLXRKKG-UHFFFAOYSA-N
1-(bromomethyl)-2-phenoxybenzene
22675469
97%
Videos
SDS