missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Bromomethyl)-3-phenoxybenzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 263.13 |
Percent Purity | 97% |
Chemical Name or Material | 1-(Bromomethyl)-3-phenoxybenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'