missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,4-dinitrobenzene, 90%, Thermo Scientific™

Catalog Number p-4229139
Click to view available options
Quantity:
50 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
610-40-2
C6H3ClN2O4
202.55
QVQSOXMXXFZAKU-UHFFFAOYSA-N
1-chloro-3,4-dinitrobenzene, 3,4-dinitrochlorobenzene, benzene, 4-chloro-1,2-dinitro, unii-z29b1f274e, 1,2-dinitro-4-chlorobenzene, 1,2-dino2 4-cl benzene, 4-chlordinitrobenzol, acmc-209mog, 4-chloro-1,2-dinitrobenzen, ghl.pd_mitscher_leg0.928
33097
4-chloro-1,2-dinitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]
This item is not returnable. View return policy
610-40-2
C6H3ClN2O4
202.55
QVQSOXMXXFZAKU-UHFFFAOYSA-N
1-chloro-3,4-dinitrobenzene, 3,4-dinitrochlorobenzene, benzene, 4-chloro-1,2-dinitro, unii-z29b1f274e, 1,2-dinitro-4-chlorobenzene, 1,2-dino2 4-cl benzene, 4-chlordinitrobenzol, acmc-209mog, 4-chloro-1,2-dinitrobenzen, ghl.pd_mitscher_leg0.928
33097
4-chloro-1,2-dinitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]
CAS Min % 88.0
CAS Max % 100.0
Melting Point 36.0°C
Color Brown-Yellow
Density 1.6800g/mL
Boiling Point 315.0°C
Flash Point 194°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(NO2)2
Refractive Index 1.5865 to 1.5885 (20°C, 589nm)
Quantity 50 g
Beilstein 05,262
Specific Gravity 1.68
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 202.55
Percent Purity 90%
Physical Form Oily Liquid After Melting
Chemical Name or Material 1-Chloro-3,4-dinitrobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-3,4-dinitrobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'