missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol, 95%, Thermo Scientific Chemicals

Catalog Number p-4510341
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
1569-02-4
C5H12O2
104.15
MFCD00067050
JOLQKTGDSGKSKJ-UHFFFAOYNA-N
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
15287
1-ethoxypropan-2-ol
CCOCC(C)O
This item is not returnable. View return policy
1569-02-4
C5H12O2
104.15
MFCD00067050
JOLQKTGDSGKSKJ-UHFFFAOYNA-N
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
15287
1-ethoxypropan-2-ol
CCOCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -100°C
Color Colorless
Density 0.8970g/mL
Boiling Point 132°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. Remark: balance 2-ethoxy-1-propanol (GC)
Packaging Glass Bottle
Linear Formula C2H5OCH2CH(OH)CH3
Refractive Index 1.4050 to 1.409
Quantity 1 L
Specific Gravity 0.897
Solubility Information Solubility in water: soluble
Viscosity 2 mPa.s (20°C)
Formula Weight 104.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Ethoxy-2-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethoxy-2-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'