missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-2,3-dimethylimidazolium chloride, 97%

Brand:  Alfa Aesar™ H59225.18

Code : NEW

Additional Details : CAS Number : 92507-97-6 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 15475468

  • 739.00 € / 50g

Estimated Shipment date
from Supplier 18-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is used reaction media for chemical processes.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

92507-97-6
160.645
NUJYCYLTUXYHQU-UHFFFAOYSA-M
2734165
CCN1C=C[N+](=C1C)C.[Cl-]
C7H13ClN2
MFCD03093290
1-ethyl-2,3-dimethylimidazolium chloride, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium chloride, ksc496m4d, 1,2-dimethyl-3-ethylimidazolium chloride, 1-ethyl-2,3-dimethyl-imidazolium chloride, 1-ethyl-2,3-dimethylimidazol-3-ium chloride, 1-ethyl-2,3-dimethylimidazol-1-ium chloride, 3-ethyl-1,2-dimethylimidazol-1-ium chloride, 1-ethyl-2,3-dimethylimidazolium chloride t
1-ethyl-2,3-dimethylimidazol-3-ium;chloride

Specifications

1-Ethyl-2,3-dimethylimidazolium chloride
50g
C7H13ClN2
1-ethyl-2,3-dimethylimidazolium chloride, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium chloride, ksc496m4d, 1,2-dimethyl-3-ethylimidazolium chloride, 1-ethyl-2,3-dimethyl-imidazolium chloride, 1-ethyl-2,3-dimethylimidazol-3-ium chloride, 1-ethyl-2,3-dimethylimidazol-1-ium chloride, 3-ethyl-1,2-dimethylimidazol-1-ium chloride, 1-ethyl-2,3-dimethylimidazolium chloride t
CCN1C=C[N+](=C1C)C.[Cl-]
160.645
160.64
97%
Moisture Sensitive
92507-97-6
MFCD03093290
NUJYCYLTUXYHQU-UHFFFAOYSA-M
1-ethyl-2,3-dimethylimidazol-3-ium;chloride
2734165
Solid
Videos
SDS