missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate, 98%, Thermo Scientific™

Catalog Number 15465086
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
307492-75-7
C7H13BF4N2
212.00
MFCD08458929
KNIWGPCMWBGRBE-UHFFFAOYSA-N
1-ethyl-2,3-dimethylimidazolium tetrafluoroborate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium tetrafluoroborate, 3-ethyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, dsstox_cid_29122, dsstox_rid_83341, dsstox_gsid_49266, 1-ethyl 2,3 dimethyl imidazolium tetrafluoroborate, 1-ethyl-2,3-dimethylimidazol-1-ium tetrafluoroborate
21977262
1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
F[B-](F)(F)F.CCN1C=C[N+](C)=C1C
This item is not returnable. View return policy
307492-75-7
C7H13BF4N2
212.00
MFCD08458929
KNIWGPCMWBGRBE-UHFFFAOYSA-N
1-ethyl-2,3-dimethylimidazolium tetrafluoroborate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium tetrafluoroborate, 3-ethyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, dsstox_cid_29122, dsstox_rid_83341, dsstox_gsid_49266, 1-ethyl 2,3 dimethyl imidazolium tetrafluoroborate, 1-ethyl-2,3-dimethylimidazol-1-ium tetrafluoroborate
21977262
1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
F[B-](F)(F)F.CCN1C=C[N+](C)=C1C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 212
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'