missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-3-methylimidazolium acetate, 97%

Brand:  Alfa Aesar™ H59277.06

Code : NEW

Additional Details : CAS Number : 143314-17-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15475538

  • 103.70 € / 5g

Estimated Shipment date
from Supplier 27-08-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is used as a solvent and as pharmaceutical intermediate.

Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

143314-17-4
170.212
XIYUIMLQTKODPS-UHFFFAOYSA-M
11658353
CCN1C=C[N+](=C1)C.CC(=O)[O-]
C8H14N2O2
MFCD06798186
1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
1-ethyl-3-methylimidazol-3-ium;acetate

Specifications

1-Ethyl-3-methylimidazolium acetate
n20/D 1.502
-20°C
143314-17-4
MFCD06798186
XIYUIMLQTKODPS-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;acetate
11658353
Liquid
1.027g/mL
Moisture Sensitive
5g
C8H14N2O2
1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
CCN1C=C[N+](=C1)C.CC(=O)[O-]
170.212
170.21
97%
Videos
SDS Documents