missing translation for 'onlineSavingsMsg'
Learn More

1-Hydrazinophthalazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. p-7026781
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11416166

Brand: Thermo Scientific Alfa Aesar B22995.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A monoamine oxidase inhibitor1-Hydrazinophthalazine hydrochloride is a non-nucleoside analog that inhibits DNA methylation and reactivates the expression of tumor suppressor genes. It acts as a non-selective MAO-A/B inhibitor and semicarbazide sensitive amine oxidase inhibitor. Further, it serves as an antihypertensive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A monoamine oxidase inhibitor1-Hydrazinophthalazine hydrochloride is a non-nucleoside analog that inhibits DNA methylation and reactivates the expression of tumor suppressor genes. It acts as a non-selective MAO-A/B inhibitor and semicarbazide sensitive amine oxidase inhibitor. Further, it serves as an antihypertensive.

Solubility
Soluble in water. Slightly soluble in diethyl ether and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-20-1
Molecular Formula C8H9ClN4
Molecular Weight (g/mol) 196.638
MDL Number MFCD00135998
InChI Key ZUXNZUWOTSUBMN-UHFFFAOYSA-N
Synonym hydralazine hydrochloride, 1-hydrazinophthalazine hydrochloride, 1-hydrazinylphthalazine hydrochloride, apresoline, aiselazine, hydralazine hcl, hydralazine chloride, hyperazin, hyperex, lopress
PubChem CID 9351
ChEBI CHEBI:31672
IUPAC Name phthalazin-1-ylhydrazine;hydrochloride
SMILES C1=CC=C2C(=C1)C=NN=C2NN.Cl

Specifications

Melting Point ∼275°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3568329
Merck Index 14,4763
Formula Weight 196.64
Percent Purity 98%
Chemical Name or Material 1-Hydrazinophthalazine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.