missing translation for 'onlineSavingsMsg'
Learn More

1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-4323194
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
827614-66-4
C13H23BN2O2
250.15
MFCD05663857
YMEBZRNYQBODKB-UHFFFAOYSA-N
1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
2759392
1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
CC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
827614-66-4
C13H23BN2O2
250.15
MFCD05663857
YMEBZRNYQBODKB-UHFFFAOYSA-N
1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
2759392
1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
CC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0030g/mL
Boiling Point 248°C to 249°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.003
Formula Weight 250.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'