Learn More
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, ACROS Organics™
Brand: Acros Organics 445790010
Code : 29
Additional Details : CAS Number : 872-50-4 Weight : 1.08000kg
Quantity | 1L |
---|
Chemical Identifiers
872-50-4 | |
99.13 | |
1-methylpyrrolidin-2-one |
C5H9NO | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
CN1CCCC1=O |
Specifications
1-Methyl-2-pyrrolidinone | |
1.67 mPa.s (20°C) | |
99.4% min. (GC) | |
15,619 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
99.13 | |
Analysis | |
Glass bottle | |
0.01% max | |
91°C | |
-24.0°C |
1.03 | |
872-50-4 | |
C5H9NO | |
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CN1CCCC1=O | |
99.13 | |
99.50% | |
1.0300g/mL | |
1.4690 to 1.4710 | |
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents | |
202.0°C | |
1L |