missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-iodo-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 10312963
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39806-90-1
MDL Number MFCD05663860
InChI Key RSDRDHPLXWMTRJ-UHFFFAOYSA-N
Synonym 4-iodo-1-methyl-1h-pyrazole, 1-methyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-methyl, 1-methyl-4-iodopyrazole, zlchem 32, pubchem24346, acmc-1ctkh, n-methyl-4-iodopyrazole, 1-methyl-4-iodo pyrazole, 4-iodo-1-methyl-pyrazole
PubChem CID 2734773
IUPAC Name 4-iodo-1-methylpyrazole
SMILES CN1C=C(C=N1)I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61°C to 67°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H5IN2
Quantity 250 mg
Molecular Weight (g/mol) 208.002
Formula Weight 208
Percent Purity ≥96.0%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material 1-Methyl-4-iodo-1H-pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-4-iodo-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'