missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-piperidinemethanol, 97%, ACROS Organics™

Brand:  Acros Organics 440480050

Code : 29

Additional Details : CAS Number : 20691-89-8 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11966401

  • 109.00 € / 5g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

20691-89-8
129.2
1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
(1-methylpiperidin-4-yl)methanol
C7H15NO
KJZLJGZZDNGGCA-UHFFFAOYSA-N
271971
CN1CCC(CC1)CO

Specifications

1-Methyl-4-piperidinemethanol
Authentic
0.98
Colorless to Yellow
5g
96.0
97%
1-methyl-4-piperidinemethanol, 1-methylpiperidin-4-yl methanol, n-methyl-4-piperidinemethanol, 4-piperidinemethanol, 1-methyl, 1-methyl-4-piperidinyl methanol, 4-hydroxymethyl-1-methylpiperidine, 1-methylpiperidine-4-methanol, n-methyl-4-hydroxymethylpiperidine, 1-methyl-piperidin-4-yl-methanol, 1-methyl-4-piperidyl methan-1-ol
CN1CCC(CC1)CO
129.2
129.2
97%
0.9800g/mL
Glass bottle
200.0°C
29.0°C to 31.0°C
20691-89-8
100.0
C7H15NO
KJZLJGZZDNGGCA-UHFFFAOYSA-N
(1-methylpiperidin-4-yl)methanol
271971
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Methyl-4-piperidinemethanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents