missing translation for 'onlineSavingsMsg'
Learn More

1-(Methylsulfonyl)-4-nitrobenzene, 95%, Thermo Scientific™

Catalog Number p-2694522
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
2976-30-9
C7H7NO4S
201.196
MFCD00047803
XONGBDXIFQIQBN-UHFFFAOYSA-N
1-methylsulfonyl-4-nitrobenzene, methyl 4-nitrophenyl sulfone, sulfone, methyl p-nitrophenyl, 1-methanesulfonyl-4-nitrobenzene, 4-methylsulfonylnitrobenzene, methyl p-nitrophenyl sulfone, 4-methylsulfonyl nitrobenzene, benzene, 1-methylsulfonyl-4-nitro, 4-methylsulfonyl nitro benzene, 1-methylsulphonyl-4-nitrobenzene
258434
1-methylsulfonyl-4-nitrobenzene
CS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
2976-30-9
C7H7NO4S
201.196
MFCD00047803
XONGBDXIFQIQBN-UHFFFAOYSA-N
1-methylsulfonyl-4-nitrobenzene, methyl 4-nitrophenyl sulfone, sulfone, methyl p-nitrophenyl, 1-methanesulfonyl-4-nitrobenzene, 4-methylsulfonylnitrobenzene, methyl p-nitrophenyl sulfone, 4-methylsulfonyl nitrobenzene, benzene, 1-methylsulfonyl-4-nitro, 4-methylsulfonyl nitro benzene, 1-methylsulphonyl-4-nitrobenzene
258434
1-methylsulfonyl-4-nitrobenzene
CS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]
CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CH3SO2C6H4NO2
Quantity 1 g
Formula Weight 201.20
Percent Purity 97%
Chemical Name or Material 1-(Methylsulfonyl)-4-nitrobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'