missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium iodide, 98%, Thermo Scientific Chemicals

Catalog Number p-7053260
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
178631-05-5
C10H19IN2
294.18
MFCD12964976
CZIUVCSYOGFUPH-UHFFFAOYSA-M
1-hexyl-3-methylimidazolium iodide, 1-hexyl-3-methyl-1h-imidazol-3-ium iodide, 1-n-hexyl-3-methylimidazolium iodide, dsstox_cid_27927, dsstox_rid_82680, dsstox_gsid_47951, 1-methyl-3-hexylimidazolium iodide, 1-hexyl-3-me-thylimidazolium iodide, 1h-imidazolium,1-hexyl-3-methyl-,iodide, 1-hexyl-3-methylimidazolium iodide hplc
12036651
1-hexyl-3-methylimidazol-3-ium;iodide
CCCCCCN1C=C[N+](=C1)C.[I-]
This item is not returnable. View return policy
178631-05-5
C10H19IN2
294.18
MFCD12964976
CZIUVCSYOGFUPH-UHFFFAOYSA-M
1-hexyl-3-methylimidazolium iodide, 1-hexyl-3-methyl-1h-imidazol-3-ium iodide, 1-n-hexyl-3-methylimidazolium iodide, dsstox_cid_27927, dsstox_rid_82680, dsstox_gsid_47951, 1-methyl-3-hexylimidazolium iodide, 1-hexyl-3-me-thylimidazolium iodide, 1h-imidazolium,1-hexyl-3-methyl-,iodide, 1-hexyl-3-methylimidazolium iodide hplc
12036651
1-hexyl-3-methylimidazol-3-ium;iodide
CCCCCCN1C=C[N+](=C1)C.[I-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 294.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'