missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium trifluoromethanesulfonate, 95%, Thermo Scientific™

Catalog Number 15474886
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
460345-16-8
C11H19F3N2O3S
316.339
MFCD03427621
RABFGPMWVQNDHI-UHFFFAOYSA-M
1-hexyl-3-methylimidazolium trifluoromethanesulfonate, 1-hexyl-3-methylimidazolium trifluoromethansulfonate, 1-hexyl-3-methylimidazolium triflate, hmim otf, dsstox_cid_27847, dsstox_rid_82600, dsstox_gsid_47871, ksc491c7h, 1-hexyl-3-methylimidazoliumtrifluoromethansulfonate, 1-hexyl-3-methyl-imidazol-3-ium; trifluoromethanesulfonate
2734247
1-hexyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]
This item is not returnable. View return policy
460345-16-8
C11H19F3N2O3S
316.339
MFCD03427621
RABFGPMWVQNDHI-UHFFFAOYSA-M
1-hexyl-3-methylimidazolium trifluoromethanesulfonate, 1-hexyl-3-methylimidazolium trifluoromethansulfonate, 1-hexyl-3-methylimidazolium triflate, hmim otf, dsstox_cid_27847, dsstox_rid_82600, dsstox_gsid_47871, ksc491c7h, 1-hexyl-3-methylimidazoliumtrifluoromethansulfonate, 1-hexyl-3-methyl-imidazol-3-ium; trifluoromethanesulfonate
2734247
1-hexyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
CCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 316.34
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium trifluoromethanesulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'