missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
Specifications
Absorbance | (0.25M aqueous solution), (0.25 M aqueous solution) at 0nm, 0.04 max. at 260nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.075 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.3 max. at 200nm |
Melting Point | >300.0°C |
Color | White |
Infrared Spectrum | Authentic |
Linear Formula | CH3(CH2)7SO3Na |
Quantity | 500 g |
Solubility Information | Solubility in water: freely soluble. Other solubilities: 4g/L in methanol |
IUPAC Name | sodium octane-1-sulfonate |
Formula Weight | 216.28 |
Percent Purity | ≥98.0% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'