missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-3598248
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -16.0°C
Color Colorless
Density 0.8240g/mL
Boiling Point 195.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7OH
Refractive Index 1.4280 to 1.4310
Quantity 250 mL
Beilstein 01, 418
Merck Index 15, 6840
Specific Gravity 0.824
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,and mineral oil,soluble in propylene glycol,insoluble in glycerol
Viscosity 9 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'