missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt hydrate, 99%, ACROS Organics™

Brand:  Acros Organics 270370250

Code : 29

Additional Details : CAS Number : 22767-49-3 Weight : 0.07500kg

Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10023230

  • 146.00 € / 25g

In stock, ready to ship: 6

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1266615-65-9
174.12
n-Amylsulfonic acid, sodium salt
C5H11NaO3S·xH2O
MFCD00149548

Specifications

(10% aq. soln.) clear, colorless
Authentic
Glass bottle
>300.0°C
1266615-65-9
C5H11NaO3S·xH2O
MFCD00149548
n-Amylsulfonic acid, sodium salt
174.12
Crystalline Powder
1-Pentanesulfonic acid, sodium salt hydrate
9.6% max. (100°C, 3 hrs) (vacuum dried)
White
25g
99%
CH3(CH2)4SO3Na·xH2O
04, III, 23
(10% in water) Clear colorless
174.12
99%
Videos
SDS Documents