missing translation for 'onlineSavingsMsg'
Learn More

1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4228881
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
14092-04-7, 109-99-9
C3H5BrMg
145.28
MFCD00209610
HHNLJRLNDSEYOX-UHFFFAOYSA-M
1-propenylmagnesium bromide, propenylmagnesium bromide, 1-propenyl magnesium bromide, magnesium, bromo-1z-1-propenyl, e-prop-1-en-1-ylmagnesium bromide, propenyl magnesium bromide, z-propenylmagnesium bromide, prop-1-enylmagnesium bromide, e-propenylmagnesium bromide, prop-1-en-1-ylmagnesium bromide, grignard reagent
5112801
magnesium;prop-1-ene;bromide
C\C=C\[Mg]Br
14092-04-7, 109-99-9
C3H5BrMg
145.28
MFCD00209610
HHNLJRLNDSEYOX-UHFFFAOYSA-M
1-propenylmagnesium bromide, propenylmagnesium bromide, 1-propenyl magnesium bromide, magnesium, bromo-1z-1-propenyl, e-prop-1-en-1-ylmagnesium bromide, propenyl magnesium bromide, z-propenylmagnesium bromide, prop-1-enylmagnesium bromide, e-propenylmagnesium bromide, prop-1-en-1-ylmagnesium bromide, grignard reagent
5112801
magnesium;prop-1-ene;bromide
C\C=C\[Mg]Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 6.88, 91.59
CAS Max % 8.41, 93.12
Color Brown to Yellow
Density 0.9500g/mL
Boiling Point 65.0°C
Flash Point −20°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.95
Solubility Information Solubility in water: reacts.
Formula Weight 145.28
Physical Form Solution
Chemical Name or Material 1-Propenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'