Learn More
Greiner Bio-One 10 mL Sterile Serological Pipets
GREENER_CHOICE
Product Code. 10043353
Click to view available options
Packaging Type:
Individual Plastic Wrap
Unit Size:
Pack of 200
Product Code. 10043353
Brand: Greiner Bio-One 607160
Only null left
Add to basket
Edge
Add to basket
Drip-free pipetting design with high optical clarity and maximum accuracy.
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Description Shape: Standard
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 1/10
Packaging Type Individual Plastic Wrap
Volume (Metric) 10 mL
Type Serological Pipet