missing translation for 'onlineSavingsMsg'
Learn More

KGW Isotherm™ Dewar Cylindrical Insulated Flasks

For the storage and insulation of temperature-sensitive goods such as dry ice, liquid nitrogen and other coolants. KGW Isotherm™ Dewar Cylindrical Insulated Flasks feature a robust design made from borosilicate glass 3.3.

Brand:  KGW Isotherm™ 10217

Additional Details : Weight : 1.20000kg

 View more versions of this product

Product Code. 10790232

  • 221.21 € / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Easy handling
  • Cooling of small samples
  • Fully silvered as standard
  • Flexible applications in laboratory
  • Available with or without side grip
Specifications

Specifications

No side grip
Spring Clips
240mm
Blue
Borosilicate Glass
100 mm
Cooling of small samples, storage of liquid nitrogen, dry ice, other coolants and as insulating flask
300mm
130mm
1500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers