missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 1000mL Media Bottle, Clear Type I

Manufactured from borosilicate glass which provides the best chemical resistance. Wheaton 1000mL Media Bottle, Clear Type I are available with black phenolic Polyethylene or Rubber lined caps.

Brand:  DWK Life Sciences W219500

Code : NEW

Additional Details : Weight : 8.61640kg

 View more versions of this product

Product Code. 15397147

  • 153.00 € / Pack of 12


Call for latest local availability
01 885 5854
Add to basket

Description

Description

  • Clear borosilicate glass bottles, non-graduated
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Hazardous communication label included for each case for content ID and safety notification
  • Tamper evident seal keeps lidded bottles sealed to protect contents from accidental misuse
Specifications

Specifications

1000mL
PE
Pour lip
Partitioned case
Media Bottle
Black
103mm
Screw
Phenolic
Clear
TYPE 1 BOROSILICATE
12 EA
Yes
38-430
231mm
Videos