missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, BAKER ANALYZED™ ACS, J.T. Baker™
87.21 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Melting Point | -84°C |
Color | Max. 15 (APHA) |
Assay Percent Range | min. 99% |
Appearance | Clear |
Density | 0.801 g/cm³ (20°C) |
Boiling Point | 117.4°C (1013 hPa) |
Quantity | 1 L |
Flash Point | 16°C |
Packaging | Glass Bottle |
Residue after Evaporation | max. 0.002% |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'