missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18 Antibody (SS18.SSX/12628R), Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-45064
This item is not returnable.
View return policy
Description
SS18 Monoclonal antibody specifically detects SS18 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| SS18 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 2-4ug/mL, Immunohistochemistry : 1-2ug/mL | |
| MGC116875, Protein SYT, SSXTsynovial sarcoma, translocated to X chromosome, Synovial sarcoma translocated to X chromosome protein, synovial sarcoma translocation, chromosome 18, SYTprotein SSXT | |
| Recombinant fragment corresponding to the fusion site of human SS18 protein (Uniprot: Q15532). Exact sequence is proprietary. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6760 | |
| Store at -20 to -84°C. Avoid freeze-thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| SS18.SSX/12628R | |
| Unconjugated | |
| 10 mM PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction