missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRA-POINT™ Solvent G for two-component system, J.T. Baker™
22.92 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
142.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "21637" to get your promotional price.
142.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
- Volumetric reagents for water determination by Karl Fischer titration
- No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
- They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
- Formulated to reach each endpoint quickly, accurately and with reproducibility
- Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
- To be used with HYDRA-POINT™ Titrant 2 product no. 8845 or HYDRA-POINT™ Titrant 5 product no. 8844
Specifications
Quantity | 1 L |
Use Test | Suitability for Karl Fischer titration: Passes Test |
UN Number | 1992 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'