missing translation for 'onlineSavingsMsg'
Learn More

11-Aminoundecanoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-58810
Click to view available options
Quantity:
5 g
100 g
250 g
This item is not returnable. View return policy
2432-99-7
C11H23NO2
201.31
MFCD00008150
GUOSQNAUYHMCRU-UHFFFAOYSA-N
undecanoic acid, 11-amino, 11-aminoundecylic acid, 11-amino-undecanoic acid, omega-aminoundecanoic acid, ccris 39, unii-sfu34976hb, rilsan t, aminoundecanoicacid, reilsan es natural, 11-amino-undecanoicaci
17083
CHEBI:82387
11-aminoundecanoic acid
C(CCCCCN)CCCCC(=O)O
This item is not returnable. View return policy
2432-99-7
C11H23NO2
201.31
MFCD00008150
GUOSQNAUYHMCRU-UHFFFAOYSA-N
undecanoic acid, 11-amino, 11-aminoundecylic acid, 11-amino-undecanoic acid, omega-aminoundecanoic acid, ccris 39, unii-sfu34976hb, rilsan t, aminoundecanoicacid, reilsan es natural, 11-amino-undecanoicaci
17083
CHEBI:82387
11-aminoundecanoic acid
C(CCCCCN)CCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 187°C to 190°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)10CO2H
Quantity 5 g
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in butanol
Formula Weight 201.31
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 11-Aminoundecanoic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'