Learn More
Hanna Instruments™ Chemical Test Kit Replacement Reagents
Pre-made reagents prepared with high-purity chemicals for ease of use. Hanna Instruments™ Chemical Test Kit Replacement Reagents are pre-measured for fast and accurate titrations using the appropriate Hanna Chemical Test Kit.
Brand:  Hanna Instruments™ HI3835100
Code : 04
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11652390
  • 157.00 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
These high quality reagents are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability. Replacement titration reagent sets provide significant cost savings compared to purchasing an entirely new Test Kit.
Chemical Test Kit Replacement Reagents are available for the following Chemical Test Kits:
  • Acidity Test Kit
  • Alkalinity Test Kit
  • Ascorbic Acid Test Kit
  • Cardon Dioxide Test Kit
  • Chloride Test Kit
  • Dissolved Oxgen Test Kit
  • Free and Total Chlorine (Low, Medium, High Range) Test Kit
  • Free Chlorine Test Kit
  • Total Chlorine Test Kit
  • Iron (Medium Range) Test Kit
  • Phosphate Test Kit
  • Salinity Test Kit
  • Sulfite Test Kit
  • Total Hardness Test Kit
Specifications
Specifications
Mercuric nitrate
HI 3835 Salinity Chemical Test Kit
110
0.0 to 40.0g/kg (ppt)
Reagent Set
Salinity determination
Bottles with Protective Cap and Instructions
1 Set
Titration